Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDC42 SIRNA Assays ›
          Gene Symbol:
          CDC42
          Gene Name:
          cell division cycle 42
          Gene Aliases:
          CDC42Hs, G25K, TKS
          Chromosome Location:
          Chr.1: 22052627 - 22092943 on Build GRCh38
          UniGene ID:
          Hs.467637
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s2765

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CDC42
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001039802.1 NP_001034891.1 3 333
          NM_001791.3 NP_001782.1 2 207
          NM_044472.2 NP_426359.1 2 207
          GenBank mRNA AF498962.1 - 1 41
          AF498963.1 - 1 41
          AK297465.1 - 1 114
          AK303994.1 - 2 123
          AK311375.1 - 2 125
          AK316449.1 - 2 124
          AL121734.1 - 2 145
          AL121735.1 - 2 145
          AL121736.1 - 3 202
          AL121737.1 - 2 276
          BC002711.2 - 2 145 3626647
          BC003682.1 - 1 94 3457085
          BC018266.1 - 3 292 3449526
          BP871988.1 - 2 207
          M35543.1 - 1 44
          M57298.1 - 2 110

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_171994, XM_008764286, XM_008764287, FQ231846, FQ232179, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001243769, NM_009861, AK151726, AK153564, AK167609, and others.

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          sprouting angiogenesis
          cardiac conduction system development
          keratinocyte development
          Golgi organization
          regulation of mitotic nuclear division
          nuclear migration
          establishment or maintenance of cell polarity
          small GTPase mediated signal transduction
          blood coagulation
          positive regulation of gene expression
          negative regulation of gene expression
          protein ubiquitination
          substantia nigra development
          actin cytoskeleton organization
          macrophage differentiation
          positive regulation of cell growth
          hair follicle morphogenesis
          positive regulation of pseudopodium assembly
          T cell costimulation
          negative regulation of protein complex assembly
          keratinization
          regulation of protein stability
          positive regulation of cytokinesis
          positive regulation of peptidyl-serine phosphorylation
          adherens junction organization
          cellular protein localization
          establishment or maintenance of apical/basal cell polarity
          multicellular organism growth
          dendritic cell migration
          Fc-gamma receptor signaling pathway involved in phagocytosis
          negative regulation of epidermal growth factor receptor signaling pathway
          regulation of protein catabolic process
          regulation of protein heterodimerization activity
          positive regulation of neuron apoptotic process
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of DNA replication
          regulation of protein kinase activity
          positive regulation of JNK cascade
          filopodium assembly
          vascular endothelial growth factor receptor signaling pathway
          ephrin receptor signaling pathway
          neuron fate determination
          actin filament bundle assembly
          regulation of small GTPase mediated signal transduction
          positive regulation of muscle cell differentiation
          regulation of filopodium assembly
          establishment of Golgi localization
          positive regulation of synapse structural plasticity
          regulation of attachment of spindle microtubules to kinetochore
          heart contraction
          canonical Wnt signaling pathway
          Wnt signaling pathway, planar cell polarity pathway
          positive regulation of epithelial cell proliferation involved in lung morphogenesis
          submandibular salivary gland formation
          epithelial-mesenchymal cell signaling
          hair follicle placode formation
          positive regulation of hair follicle cell proliferation
          organelle transport along microtubule
          actin filament branching
          epithelial cell-cell adhesion
          positive regulation of intracellular protein transport
          positive regulation of substrate adhesion-dependent cell spreading
          GTPase activity
          protein binding
          GTP binding
          protein kinase binding
          GTP-dependent protein binding
          mitogen-activated protein kinase kinase kinase binding
          thioesterase binding
          apolipoprotein A-I receptor binding
          identical protein binding
          Rho GDP-dissociation inhibitor binding
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0