Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PLCE1 SIRNA Assays ›
          Gene Symbol:
          PLCE1
          Gene Name:
          phospholipase C epsilon 1
          Gene Aliases:
          NPHS3, PLCE, PPLC
          Chromosome Location:
          Chr.10: 93993989 - 94329447 on Build GRCh38
          UniGene ID:
          Hs.655033
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s27660
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PLCE1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001165979.2 NP_001159451.1 2 529
          NM_001288989.1 NP_001275918.1 3 2018
          NM_016341.3 NP_057425.3 3 2018
          XM_006717885.3 XP_006717948.1 3 1966
          XM_006717888.3 XP_006717951.1 3 1966
          XM_006717889.3 XP_006717952.1 3 1966
          XM_006717890.2 XP_006717953.1 2 482
          XM_011539849.2 XP_011538151.1 3 3123
          XM_011539850.2 XP_011538152.1 2 1611
          XM_011539851.2 XP_011538153.1 3 1968
          XM_011539852.2 XP_011538154.1 3 1968
          XM_017016310.1 XP_016871799.1 4 2013
          XM_017016311.1 XP_016871800.1 3 1966
          XM_017016312.1 XP_016871801.1 2 482
          GenBank mRNA AB040949.2 - 3 2018
          AF170071.1 - 2 460
          AF190642.2 - 2 1619
          AK289852.1 - 2 482
          AK297779.1 - 2 269
          BC140705.1 - 2 1444 9021573
          BC144286.1 - 2 1444 9052806
          BC151854.1 - 3 2018 8860521

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00275279_m1

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor phospholipase

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          regulation of cell growth
          phospholipid metabolic process
          diacylglycerol biosynthetic process
          regulation of smooth muscle contraction
          cytoskeleton organization
          epidermal growth factor receptor signaling pathway
          phospholipase C-activating G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          protein kinase C-activating G-protein coupled receptor signaling pathway
          Ras protein signal transduction
          heart development
          regulation of G-protein coupled receptor protein signaling pathway
          cell proliferation
          lipid catabolic process
          calcium-mediated signaling
          glomerulus development
          positive regulation of GTPase activity
          inositol phosphate metabolic process
          regulation of protein kinase activity
          regulation of Ras protein signal transduction
          inositol phosphate-mediated signaling
          phosphatidylinositol phospholipase C activity
          phospholipase C activity
          receptor signaling protein activity
          guanyl-nucleotide exchange factor activity
          protein binding
          Ras GTPase binding
          enzyme binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0