Search
Search

ECSIT| Gene Symbol: ECSIT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142464.2 | NP_001135936.1 | 6 | 1025 | ||
| NM_001142465.2 | NP_001135937.1 | 5 | 532 | |||
| NM_001243204.1 | NP_001230133.1 | 7 | 1063 | |||
| NM_016581.4 | NP_057665.2 | 7 | 1174 | |||
| GenBank mRNA | AF243044.1 | - | 6 | 1073 | ||
| AK314905.1 | - | 7 | 1063 | |||
| BC000193.1 | - | 7 | 1122 | 3352568 | ||
| BC005119.1 | - | 7 | 1117 | 2819610 | ||
| BC008279.1 | - | 6 | 990 | 3639801 | ||
| BE791936.1 | - | 5 | 533 | |||
| BU186092.1 | - | 4 | 305 | |||
| CR457204.1 | - | 6 | 1040 | |||