Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDK5 SIRNA Assays ›
          Gene Symbol:
          CDK5
          Gene Name:
          cyclin dependent kinase 5
          Gene Aliases:
          LIS7, PSSALRE
          Chromosome Location:
          Chr.7: 151053812 - 151057965 on Build GRCh38
          UniGene ID:
          Hs.647078
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s2826
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CDK5
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164410.2 NP_001157882.1
          5 416
          NM_004935.3 NP_004926.1
          5 416
          GenBank mRNA AK026533.1 -
          5 337
          AY049778.1 -
          5 299
          AY927560.1 - LIT3563
          AY927560
          1 125
          BC005115.1 -
          5 324 3537202
          BP309068.1 -
          5 396
          BT006680.1 -
          5 299
          BU527089.1 -
          5 364
          CB114175.1 -
          5 342
          CR457041.1 -
          5 299
          DB487008.1 -
          5 416
          DQ411039.1 -
          5 299
          KU177968.1 -
          5 299
          KU177969.1 -
          5 299
          L04658.1 -
          5 311
          X66364.1 -
          5 323

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00358991_g1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          synaptic transmission, dopaminergic
          intracellular protein transport
          nucleocytoplasmic transport
          cell cycle
          cell-matrix adhesion
          chemical synaptic transmission
          synapse assembly
          skeletal muscle tissue development
          motor neuron axon guidance
          cell proliferation
          visual learning
          response to wounding
          Schwann cell development
          synaptic vesicle exocytosis
          regulation of macroautophagy
          phosphorylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          sensory perception of pain
          cerebellar cortex formation
          hippocampus development
          layer formation in cerebral cortex
          central nervous system neuron development
          corpus callosum development
          neuron differentiation
          regulation of cell migration
          negative regulation of axon extension
          cortical actin cytoskeleton organization
          neuron projection development
          negative regulation of protein ubiquitination
          negative regulation of synaptic plasticity
          positive regulation of protein binding
          receptor catabolic process
          serine phosphorylation of STAT3 protein
          synaptic transmission, glutamatergic
          protein localization to synapse
          regulation of apoptotic process
          receptor clustering
          positive regulation of neuron apoptotic process
          negative regulation of cell cycle
          positive regulation of protein kinase activity
          negative regulation of proteolysis
          negative regulation of transcription, DNA-templated
          positive regulation of calcium ion-dependent exocytosis
          protein autophosphorylation
          negative regulation of protein export from nucleus
          behavioral response to cocaine
          regulation of synaptic plasticity
          synaptic vesicle endocytosis
          rhythmic process
          axon extension
          oligodendrocyte differentiation
          dendrite morphogenesis
          cell division
          neuron apoptotic process
          excitatory postsynaptic potential
          regulation of dendritic spine morphogenesis
          calcium ion import
          regulation of cell cycle arrest
          positive regulation of protein targeting to membrane
          negative regulation of neuron death
          regulation of signal transduction by p53 class mediator
          regulation of synaptic vesicle recycling
          positive regulation of actin cytoskeleton reorganization
          positive regulation of receptor activity
          p53 binding
          protein kinase activity
          protein serine/threonine kinase activity
          cyclin-dependent protein serine/threonine kinase activity
          ErbB-2 class receptor binding
          protein binding
          ATP binding
          cytoskeletal protein binding
          kinase activity
          protein kinase binding
          acetylcholine receptor activator activity
          ErbB-3 class receptor binding
          ephrin receptor binding
          tau-protein kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0