Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MED15 SIRNA Assays ›
          Gene Symbol:
          MED15
          Gene Name:
          mediator complex subunit 15
          Gene Aliases:
          ARC105, CAG7A, CTG7A, PCQAP, TIG-1, TIG1, TNRC7
          Chromosome Location:
          Chr.22: 20507542 - 20587632 on Build GRCh38
          UniGene ID:
          Hs.517421
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s28367

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MED15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001003891.2 NP_001003891.1 12, 13 1804
          NM_001293234.1 NP_001280163.1 11, 12 1633
          NM_001293235.1 NP_001280164.1 11, 12 1673
          NM_001293236.1 NP_001280165.1 10, 11 1471
          NM_001293237.1 NP_001280166.1 11, 12 1876
          NM_015889.4 NP_056973.2 11, 12 1684
          XM_006724263.3 XP_006724326.1 11, 12 1549
          XM_006724264.3 XP_006724327.1 12, 13 1775
          XM_011530214.2 XP_011528516.1 13, 14 1848
          XM_011530215.2 XP_011528517.1 11, 12 1670
          XM_017028817.1 XP_016884306.1 11, 12 1746
          XM_017028818.1 XP_016884307.1 13, 14 1946
          XM_017028819.1 XP_016884308.1 12, 13 1712
          XM_017028820.1 XP_016884309.1 11, 12 1655
          XM_017028823.1 XP_016884312.1 11, 12 2927
          XM_017028824.1 XP_016884313.1 13, 14 1844
          GenBank mRNA AF056191.1 - 11, 12 1633
          AF328769.1 - 11, 12 1541
          AK000003.1 - 1, 2 1149
          AK074230.1 - 6, 7 831
          AK074268.1 - 11, 12 1478
          AK090465.1 - 12, 13 1736
          AK124276.1 - 4, 5 1950
          AK124925.1 - 4, 5 1155
          AK126208.1 - 7, 8 2009
          AK294540.1 - 13, 14 1680
          AK294734.1 - 11, 12 1872
          AK297284.1 - 6, 7 1062
          AK315540.1 - 11, 12 1596
          AL832151.1 - 4, 5 819
          AL832923.1 - 1, 2 137
          BC003078.1 - 11, 12 1605
          BC007529.1 - 4, 5 457
          BC013985.2 - 11, 12 1587 3506327
          BC017110.1 - 8, 9 1432
          CR456537.1 - 10, 11 1363
          U80745.1 - 6, 7 995

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          RNA polymerase II transcription cofactor activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0