Search
Search

HSPB11| Gene Symbol: HSPB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316935.1 | NP_001303864.1 | 2, 3 | 185 | ||
| NM_016126.3 | NP_057210.2 | 2, 3 | 382 | |||
| NR_133633.1 | 2, 3 | 382 | ||||
| GenBank mRNA | AF100747.1 | - | 2, 3 | 155 | ||
| AF416725.1 | - | 1, 2 | 109 | |||
| AF417114.1 | - | 1, 2 | 109 | |||
| BC005245.1 | - | 2, 3 | 174 | 3838843 | ||
| BF967485.1 | - | 2, 3 | 491 | |||
| DA133366.1 | - | 2, 3 | 258 | |||
| DB480705.1 | - | 2, 3 | 382 | |||
| HY179788.1 | - | 2, 3 | 186 | |||