Search
Search

FBXO40| Gene Symbol: FBXO40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016298.3 | NP_057382.2 | 4 | 2213 | ||
| GenBank mRNA | AB033021.1 | - | 4 | 2417 | ||
| AF204674.1 | - | 4 | 2119 | |||
| AK314400.1 | - | 4 | 2135 | |||
| BC109275.1 | - | 3 | 2024 | 40028315 | ||
| BC109276.1 | - | 3 | 2024 | 40028318 | ||