Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRMT7 SIRNA Assays ›
          Gene Symbol:
          PRMT7
          Gene Name:
          protein arginine methyltransferase 7
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 68310974 - 68358832 on Build GRCh38
          UniGene ID:
          Hs.640229
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s29077

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRMT7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001184824.1 NP_001171753.1 6, 7 813
          NM_001290018.1 NP_001276947.1 8, 9 1604
          NM_019023.2 NP_061896.1 8, 9 1098
          XM_011523112.2 XP_011521414.1 7, 8 963
          XM_011523113.2 XP_011521415.1 8, 9 1090
          XM_011523115.2 XP_011521417.1 7, 8 907
          XM_011523116.2 XP_011521418.1 7, 8 963
          XM_011523121.2 XP_011521423.1 5, 6 728
          XM_011523124.2 XP_011521426.1 7, 8 1016
          XM_011523125.2 XP_011521427.1 6, 7 858
          XM_011523126.2 XP_011521428.1 6, 7 862
          XM_011523128.2 XP_011521430.1 7, 8 907
          XM_011523131.2 XP_011521433.1 6, 7 837
          XM_017023290.1 XP_016878779.1 8, 9 1090
          XM_017023291.1 XP_016878780.1 8, 9 1030
          XM_017023292.1 XP_016878781.1 7, 8 907
          XM_017023293.1 XP_016878782.1 8, 9 1090
          XM_017023294.1 XP_016878783.1 8, 9 1030
          XM_017023295.1 XP_016878784.1 7, 8 963
          XM_017023296.1 XP_016878785.1 8, 9 1090
          XM_017023297.1 XP_016878786.1 8, 9 1090
          XM_017023298.1 XP_016878787.1 8, 9 1090
          XM_017023299.1 XP_016878788.1 8, 9 1090
          XM_017023300.1 XP_016878789.1 8, 9 1090
          XM_017023301.1 XP_016878790.1 8, 9 1090
          XM_017023302.1 XP_016878791.1 6, 7 858
          XM_017023303.1 XP_016878792.1 7, 8 1016
          XM_017023304.1 XP_016878793.1 8, 9 1090
          XM_017023305.1 XP_016878794.1 5, 6 728
          XM_017023306.1 XP_016878795.1 8, 9 1090
          XM_017023307.1 XP_016878796.1 7, 8 1016
          XM_017023308.1 XP_016878797.1 7, 8 1030
          XM_017023309.1 XP_016878798.1 6, 7 858
          XM_017023310.1 XP_016878799.1 6, 7 882
          XM_017023311.1 XP_016878800.1 6, 7 814
          XM_017023312.1 XP_016878801.1 7, 8 963
          XM_017023313.1 XP_016878802.1 7, 8 1005
          XM_017023314.1 XP_016878803.1 6, 7 765
          XM_017023315.1 XP_016878804.1 7, 8 1005
          XM_017023316.1 XP_016878805.1 6, 7 765
          GenBank mRNA AB067520.1 - 2, 3 1100
          AK001502.1 - 8, 9 1030
          AK022739.1 - 4, 5 525
          AK056647.1 - 8, 9 1586
          AK097175.1 - 6, 7 1540
          AK304605.1 - 6, 7 736
          BC000146.1 - 6, 7 839 2900965

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          spliceosomal snRNP assembly
          regulation of gene expression by genetic imprinting
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          histone methylation
          peptidyl-arginine methylation
          peptidyl-arginine methylation, to symmetrical-dimethyl arginine
          peptidyl-arginine methylation, to asymmetrical-dimethyl arginine
          cell differentiation
          histone arginine methylation
          DNA methylation involved in gamete generation
          regulation of protein binding
          histone H4-R3 methylation
          histone-arginine N-methyltransferase activity
          S-adenosylmethionine-dependent methyltransferase activity
          protein-arginine N-methyltransferase activity
          [myelin basic protein]-arginine N-methyltransferase activity
          protein-arginine omega-N monomethyltransferase activity
          protein-arginine omega-N asymmetric methyltransferase activity
          protein-arginine omega-N symmetric methyltransferase activity
          histone binding
          ribonucleoprotein complex binding
          histone methyltransferase activity (H4-R3 specific)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0