Search
Search

UGT1A9| Gene Symbol: UGT1A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021027.2 | NP_066307.1 | 1 | 352 | ||
| GenBank mRNA | AF056188.1 | - | 1 | 333 | ||
| AK313510.1 | - | 1 | 391 | |||
| AY435144.1 | - | 1 | 315 | |||
| BC058844.1 | - | 1 | 358 | 4593613 | ||
| S55985.1 | - | 1 | 349 | |||