Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NDE1 SIRNA Assays ›
          Gene Symbol:
          NDE1
          Gene Name:
          nudE neurodevelopment protein 1
          Gene Aliases:
          HOM-TES-87, LIS4, MHAC, NDE, NUDE, NUDE1
          Chromosome Location:
          Chr.16: 15643267 - 15726353 on Build GRCh38
          UniGene ID:
          Hs.655378
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s29511

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NDE1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001143979.1 NP_001137451.1 5 1156
          NM_017668.2 NP_060138.1 4 442
          XM_005255396.4 XP_005255453.1 4 435
          XM_006720897.3 XP_006720960.1 4 1002
          XM_006720900.3 XP_006720963.1 4 471
          XM_011522553.2 XP_011520855.1 5 1156
          XM_017023349.1 XP_016878838.1 4 435
          XM_017023350.1 XP_016878839.1 4 425
          XM_017023351.1 XP_016878840.1 4 435
          XM_017023352.1 XP_016878841.1 4 435
          XM_017023353.1 XP_016878842.1 4 435
          XM_017023354.1 XP_016878843.1 4 435
          XM_017023355.1 XP_016878844.1 4 435
          XM_017023356.1 XP_016878845.1 4 471
          XM_017023357.1 XP_016878846.1 2 189
          GenBank mRNA AF124431.1 - 3 351
          AK000108.1 - 4 421
          AK025815.1 - 4 421
          BC001421.2 - 4 435 3140369
          BC033900.1 - 5 1161 4828494
          BP271845.1 - 4 442
          KJ534887.1 - 3 330
          KJ534967.1 - 3 330
          KJ534991.1 - 3 330

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          establishment of mitotic spindle orientation
          neuron migration
          microtubule nucleation
          chromosome segregation
          sister chromatid cohesion
          mitotic centrosome separation
          neuroblast proliferation
          cell migration
          cerebral cortex development
          vesicle transport along microtubule
          centrosome duplication
          cell division
          establishment of chromosome localization
          centrosome localization
          regulation of microtubule motor activity
          protein binding
          microtubule binding
          protein domain specific binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline