Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ST7L SIRNA Assays ›
          Gene Symbol:
          ST7L
          Gene Name:
          suppression of tumorigenicity 7 like
          Gene Aliases:
          FAM4B, ST7R, STLR
          Chromosome Location:
          Chr.1: 112517444 - 112620838 on Build GRCh38
          UniGene ID:
          Hs.201921
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s29661

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ST7L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001308264.1 NP_001295193.1 6, 7 951
          NM_017744.4 NP_060214.2 7, 8 1160
          NM_138727.3 NP_620055.1 7, 8 1109
          NM_138728.2 NP_620056.1 7, 8 1160
          NM_138729.3 NP_620057.1 7, 8 1160
          XM_005270963.4 XP_005271020.1 7, 8 683
          XM_005270964.3 XP_005271021.1 6, 7 775
          XM_011541627.2 XP_011539929.1 7, 8 869
          XM_011541628.2 XP_011539930.1 7, 8 869
          XM_011541629.2 XP_011539931.1 7, 8 883
          XM_011541630.2 XP_011539932.1 7, 8 1579
          XM_011541631.2 XP_011539933.1 7, 8 707
          XM_017001534.1 XP_016857023.1 7, 8 869
          XM_017001535.1 XP_016857024.1 7, 8 869
          XM_017001536.1 XP_016857025.1 6, 7 775
          XM_017001537.1 XP_016857026.1 5, 6 595
          GenBank mRNA AB081317.1 - 7, 8 931
          AB081318.1 - 7, 8 880
          AB081319.1 - 7, 8 931
          AB081320.1 - 7, 8 931
          AK095837.1 - 5, 6 468
          AK097837.1 - 6, 7 740
          AK291042.1 - 7, 8 861
          AK295955.1 - 5, 6 663
          AK301869.1 - 7, 8 861
          AK304004.1 - 7, 8 818
          AK308727.1 - 5, 6 705
          BC027898.1 - 6, 7 762
          BC037830.1 - 7, 8 1573 4812133
          BC047392.1 - 7, 8 886 5273549
          BC065501.1 - 7, 8 1067 4417786
          BX640667.1 - 7, 8 836

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s)

          negative regulation of cell growth

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-tlt54:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0