Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PXK SIRNA Assays ›
          Gene Symbol:
          PXK
          Gene Name:
          PX domain containing serine/threonine kinase like
          Gene Aliases:
          MONaKA
          Chromosome Location:
          Chr.3: 58332868 - 58426127 on Build GRCh38
          UniGene ID:
          Hs.190544
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s29710
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PXK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001289098.1 NP_001276027.1 1, 2 208
          NM_017771.4 NP_060241.2 1, 2 208
          XM_005265250.4 XP_005265307.1 1, 2 208
          XM_005265252.4 XP_005265309.1 1, 2 208
          XM_005265255.3 XP_005265312.1 1, 2 208
          XM_005265256.3 XP_005265313.1 1, 2 208
          XM_017006675.1 XP_016862164.1 1, 2 208
          XM_017006676.1 XP_016862165.1 1, 2 208
          XM_017006677.1 XP_016862166.1 1, 2 208
          XM_017006678.1 XP_016862167.1 1, 2 208
          XM_017006679.1 XP_016862168.1 1, 2 208
          XM_017006680.1 XP_016862169.1 1, 2 208
          XM_017006686.1 XP_016862175.1 1, 2 166
          XM_017006687.1 XP_016862176.1 1, 2 164
          XM_017006692.1 XP_016862181.1 1, 2 203
          XM_017006693.1 XP_016862182.1 1, 2 203
          XM_017006694.1 XP_016862183.1 1, 2 97
          XM_017006695.1 XP_016862184.1 1, 2 203
          GenBank mRNA AB209688.1 - 1, 2 187
          AF399753.1 - 1, 2 109
          AK000342.1 - 1, 2 206
          AY274811.1 - 1, 2 208
          AY847222.1 - 1, 2 111
          BC008943.2 - 1, 2 189
          BC014479.1 - 1, 2 124 4856139
          DA296189.1 - 1, 2 209
          DQ124707.1 - 1, 2 109
          DQ124708.1 - 1, 2 109
          KF774202.1 - 1, 2 218
          KF774203.1 - 1, 2 218

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00298958_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          inflammatory response
          negative regulation of ATPase activity
          regulation of membrane potential
          negative regulation of ion transport
          modulation of synaptic transmission
          nucleotide binding
          actin binding
          protein kinase activity
          ATP binding
          protein C-terminus binding
          phosphatidylinositol binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0