Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CHD3 SIRNA Assays ›
          Gene Symbol:
          CHD3
          Gene Name:
          chromodomain helicase DNA binding protein 3
          Gene Aliases:
          Mi-2a, Mi2-ALPHA, ZFH
          Chromosome Location:
          Chr.17: 7884764 - 7912757 on Build GRCh38
          UniGene ID:
          Hs.25601
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s2981

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CHD3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001005271.2 NP_001005271.2 26 4297
          NM_001005273.2 NP_001005273.1 26 4268
          NM_005852.3 NP_005843.2 26 4268
          XM_005256427.4 XP_005256484.1 26 4338
          XM_005256428.4 XP_005256485.1 26 4338
          XM_005256429.4 XP_005256486.1 26 4338
          XM_006721423.3 XP_006721486.1 26 4338
          XM_006721424.3 XP_006721487.1 26 4313
          XM_006721428.3 XP_006721491.1 26 4338
          XM_017024061.1 XP_016879550.1 26 4320
          XM_017024062.1 XP_016879551.1 26 4295
          XM_017024063.1 XP_016879552.1 26 4253
          XM_017024064.1 XP_016879553.1 26 4313
          XM_017024065.1 XP_016879554.1 25 4134
          XM_017024066.1 XP_016879555.1 27 4073
          XM_017024067.1 XP_016879556.1 27 4256
          XM_017024068.1 XP_016879557.1 26 4229
          XM_017024069.1 XP_016879558.1 26 4120
          XM_017024070.1 XP_016879559.1 26 4295
          GenBank mRNA AK125928.1 - 3 681
          AK296937.1 - 8 1062
          AK308362.1 - 4 428
          BC110648.1 - 2 101
          U91543.1 - 26 4268

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin assembly or disassembly
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          spindle organization
          histone deacetylation
          DNA duplex unwinding
          centrosome organization
          regulation of signal transduction by p53 class mediator
          DNA binding
          ATP-dependent DNA helicase activity
          helicase activity
          histone deacetylase activity
          protein binding
          ATP binding
          zinc ion binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0