Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256875.1 | NP_001243804.1 | 6 | 745 | ||
NM_018101.3 | NP_060571.1 | 7 | 648 | |||
GenBank mRNA | AK001330.1 | - | 7 | 568 | ||
AK022104.1 | - | 6 | 650 | |||
AK022606.1 | - | 6 | 554 | |||
AK222549.1 | - | 7 | 568 | |||
AK291917.1 | - | 6 | 600 | |||
AK302116.1 | - | 5 | 412 | |||
AY508815.2 | - | 7 | 568 | |||
BC000703.2 | - | 6 | 641 | 3347875 | ||
BC001651.2 | - | 6 | 659 | 2822261 | ||
BC008079.2 | - | 7 | 501 | 2959649 | ||
BC016944.1 | - | 7 | 521 | 3457122 |