Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HHAT SIRNA Assays ›
          Gene Symbol:
          HHAT
          Gene Name:
          hedgehog acyltransferase
          Gene Aliases:
          MART2, SKI1, Skn
          Chromosome Location:
          Chr.1: 210327873 - 210676298 on Build GRCh38
          UniGene ID:
          Hs.58650
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s31366

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HHAT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001122834.3 NP_001116306.1 3 307
          NM_001170564.2 NP_001164035.1 3 307
          NM_001170580.2 NP_001164051.1 3 362
          NM_001170587.2 NP_001164058.1 2 239
          NM_001170588.2 NP_001164059.1 3 373
          NM_018194.5 NP_060664.2 3 373
          XM_006711441.3 XP_006711504.1 4 434
          XM_011509737.2 XP_011508039.1 4 434
          XM_011509738.2 XP_011508040.1 2 192
          XM_011509739.2 XP_011508041.1 3 359
          XM_011509740.2 XP_011508042.1 4 563
          XM_011509741.2 XP_011508043.1 3 307
          XM_011509742.2 XP_011508044.1 4 411
          XM_011509746.2 XP_011508048.1 3 388
          XM_017001728.1 XP_016857217.1 3 517
          XM_017001729.1 XP_016857218.1 3 238
          XM_017001732.1 XP_016857221.1 3 198
          XM_017001735.1 XP_016857224.1 3 838
          XM_017001736.1 XP_016857225.1 2 214
          XM_017001737.1 XP_016857226.1 3 198
          XM_017001738.1 XP_016857227.1 3 305
          XM_017001739.1 XP_016857228.1 3 394
          XM_017001740.1 XP_016857229.1 3 895
          GenBank mRNA AK001586.1 - 3 327
          AK297193.1 - 3 305
          AK298991.1 - 3 327
          AK302955.1 - 2 192
          AK316190.1 - 2 193
          AK316524.1 - 3 420
          AL049848.1 - 1 40
          BC039071.1 - 3 315 5177926
          BC117130.1 - 2 162 40125681
          DA336461.1 - 3 359
          DB077466.1 - 3 297
          DB080645.1 - 3 307

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          acyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          smoothened signaling pathway
          multicellular organism development
          protein palmitoylation
          positive regulation of protein targeting to mitochondrion
          GTP binding
          O-acyltransferase activity
          palmitoyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0