Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CSGALNACT1 SIRNA Assays ›
Gene Symbol:
CSGALNACT1
Gene Name:
chondroitin sulfate N-acetylgalactosaminyltransferase 1
Gene Aliases:
CSGalNAcT-1, ChGn, beta4GalNAcT
Chromosome Location:
Chr.8: 19404161 - 19757940 on Build GRCh38
UniGene ID:
Hs.613729
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31519
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
392.00
Your Price
Online offer:
380.65
380.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CSGALNACT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130518.1
NP_001123990.1
10
2425
NM_018371.4
NP_060841.5
10
2015
NR_024040.1
10
2094
XM_006716358.2
XP_006716421.1
9
2306
XM_006716359.2
XP_006716422.1
10
2675
XM_006716360.2
XP_006716423.1
11
2823
XM_006716361.2
XP_006716424.1
10
2570
XM_006716362.2
XP_006716425.1
9
1893
XM_006716363.1
XP_006716426.1
9
2065
XM_006716364.3
XP_006716427.1
8
1749
XM_011544578.1
XP_011542880.1
12
2942
XM_011544579.1
XP_011542881.1
11
2794
XM_011544580.1
XP_011542882.1
10
2672
XM_011544582.1
XP_011542884.1
10
2680
XM_011544583.1
XP_011542885.1
8
1859
XM_011544584.1
XP_011542886.1
8
1743
XM_011544585.2
XP_011542887.1
7
826
XM_017013623.1
XP_016869112.1
13
2992
XM_017013624.1
XP_016869113.1
10
2631
XM_017013625.1
XP_016869114.1
8
1908
XM_017013626.1
XP_016869115.1
9
1838
XM_017013627.1
XP_016869116.1
10
4232
XM_017013628.1
XP_016869117.1
9
4092
XM_017013629.1
XP_016869118.1
10
2023
XM_017013630.1
XP_016869119.1
10
1936
XM_017013631.1
XP_016869120.1
9
1897
XM_017013632.1
XP_016869121.1
10
2006
XM_017013633.1
XP_016869122.1
10
1993
XM_017013634.1
XP_016869123.1
10
1990
XM_017013635.1
XP_016869124.1
10
1998
XM_017013636.1
XP_016869125.1
11
2141
XM_017013637.1
XP_016869126.1
10
1998
XM_017013638.1
XP_016869127.1
11
2183
XM_017013639.1
XP_016869128.1
9
1831
GenBank mRNA
AB071403.1
-
7
1385
AB081516.1
-
8
1703
AK002126.1
-
7
952
AK314625.1
-
9
1774
AL157483.1
-
3
222
AY358441.1
-
10
2015
BC060772.1
-
10
2107
30340553
BX649103.1
-
10
2375
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
endochondral ossification
nervous system development
cell recognition
cell proliferation
anatomical structure morphogenesis
heparan sulfate proteoglycan biosynthetic process, polysaccharide chain biosynthetic process
UDP-N-acetylgalactosamine metabolic process
proteoglycan biosynthetic process
extracellular matrix organization
chondroitin sulfate biosynthetic process
heparin biosynthetic process
UDP-glucuronate metabolic process
chondroitin sulfate proteoglycan biosynthetic process
dermatan sulfate proteoglycan biosynthetic process
chondroitin sulfate proteoglycan biosynthetic process, polysaccharide chain biosynthetic process
cartilage development
acetylgalactosaminyltransferase activity
peptidoglycan glycosyltransferase activity
glucuronosyltransferase activity
metal ion binding
glucuronylgalactosylproteoglycan 4-beta-N-acetylgalactosaminyltransferase activity
glucuronosyl-N-acetylgalactosaminyl-proteoglycan 4-beta-N-acetylgalactosaminyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline