Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIAA1217 SIRNA Assays ›
Gene Symbol:
KIAA1217
Gene Name:
KIAA1217
Gene Aliases:
SKT
Chromosome Location:
Chr.10: 23694746 - 24547848 on Build GRCh38
UniGene ID:
Hs.445885
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s32069
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIAA1217
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098500.2
NP_001091970.1
4
984
NM_001282767.1
NP_001269696.1
3
818
NM_001282768.1
NP_001269697.1
3
818
NM_019590.4
NP_062536.2
3
818
XM_011519552.2
XP_011517854.2
4
873
XM_011519555.2
XP_011517857.2
3
791
XM_011519558.2
XP_011517860.2
4
875
XM_011519559.2
XP_011517861.2
3
565
XM_011519562.2
XP_011517864.2
4
873
XM_011519564.2
XP_011517866.1
3
545
XM_011519565.2
XP_011517867.2
3
564
XM_011519566.2
XP_011517868.2
3
792
XM_017016416.1
XP_016871905.1
4
899
XM_017016417.1
XP_016871906.1
3
818
XM_017016418.1
XP_016871907.1
2
1535
XM_017016419.1
XP_016871908.1
2
1535
XM_017016420.1
XP_016871909.1
2
282
XM_017016421.1
XP_016871910.1
4
492
XM_017016422.1
XP_016871911.1
2
282
XM_017016425.1
XP_016871914.1
4
875
XM_017016426.1
XP_016871915.1
4
879
XM_017016427.1
XP_016871916.1
4
645
XM_017016428.1
XP_016871917.1
1
61
XM_017016429.1
XP_016871918.1
5
1033
GenBank mRNA
AB033043.2
-
4
984
AB291616.1
-
2
345
AK091142.1
-
3
818
AK096446.1
-
2
206
AK303905.1
-
2
330
AL833280.1
-
3
445
BC136521.1
-
3
436
9020511
BC144227.1
-
3
436
9052747
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
embryonic skeletal system development
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0