Search
Search

COX6A1| Gene Symbol: COX6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004373.3 | NP_004364.2 | 3 | 306 | ||
| GenBank mRNA | AK312009.1 | - | 3 | 306 | ||
| BC007723.2 | - | 1 | 271 | 4300921 | ||
| BC070186.1 | - | 1 | 279 | 6654310 | ||
| BC107861.1 | - | 1 | 287 | 4664958 | ||
| BP378295.1 | - | 3 | 277 | |||
| HE647860.1 | - | 3 | 269 | |||
| X15341.1 | - | 3 | 291 | |||