Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CPS1 SIRNA Assays ›
          Gene Symbol:
          CPS1
          Gene Name:
          carbamoyl-phosphate synthase 1
          Gene Aliases:
          CPSASE1, PHN
          Chromosome Location:
          Chr.2: 210477682 - 210679107 on Build GRCh38
          UniGene ID:
          Hs.149252
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s3464

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CPS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001122633.2 NP_001116105.1 15 1531
          NM_001122634.3 NP_001116106.1 4 587
          NM_001875.4 NP_001866.2 14 1566
          XM_011510640.1 XP_011508942.1 16 1765
          XM_011510641.2 XP_011508943.1 16 1909
          XM_011510642.2 XP_011508944.1 15 1781
          XM_011510643.2 XP_011508945.1 16 1646
          XM_011510644.2 XP_011508946.1 15 1687
          GenBank mRNA AB180933.1 - 14 1431
          AB180934.1 - 14 1431
          AB180935.1 - 14 1431
          AB180936.1 - 14 1431
          AB180937.1 - 14 1431
          AB180938.1 - 14 1431
          AB208800.1 - 15 1507
          AF154830.1 - 14 1554
          AK289749.1 - 14 1566
          AK302778.1 - 4 576
          AK316425.1 - 4 587
          AY317138.1 - 4 573
          BC140943.1 - 14 1533 9056755
          D90282.1 - 14 1549
          Y15793.1 - 14 1431

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          urea cycle
          glycogen catabolic process
          'de novo' pyrimidine nucleobase biosynthetic process
          proteolysis
          arginine biosynthetic process
          glutamine metabolic process
          midgut development
          response to toxic substance
          response to zinc ion
          response to amine
          citrulline biosynthetic process
          triglyceride catabolic process
          response to food
          response to lipopolysaccharide
          response to drug
          response to starvation
          response to amino acid
          cellular response to fibroblast growth factor stimulus
          positive regulation of vasodilation
          nitric oxide metabolic process
          homocysteine metabolic process
          anion homeostasis
          response to growth hormone
          hepatocyte differentiation
          carbamoyl phosphate biosynthetic process
          cellular response to cAMP
          cellular response to glucagon stimulus
          cellular response to oleic acid
          response to dexamethasone
          aspartate carbamoyltransferase activity
          carbamoyl-phosphate synthase (ammonia) activity
          carbamoyl-phosphate synthase (glutamine-hydrolyzing) activity
          dihydroorotase activity
          endopeptidase activity
          calcium ion binding
          protein binding
          ATP binding
          phospholipid binding
          glutamate binding
          protein complex binding
          modified amino acid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0