Search
Search

IPPK| Gene Symbol: IPPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022755.5 | NP_073592.1 | 13 | 1652 | ||
| XM_017015041.1 | XP_016870530.1 | 14 | 1722 | |||
| GenBank mRNA | AF351202.1 | - | 13 | 1458 | ||
| AF520811.1 | - | 13 | 1375 | |||
| AK023225.1 | - | 13 | 1442 | |||
| AK024267.1 | - | 11 | 1230 | |||
| AK096392.1 | - | 2 | 970 | |||
| AK123694.1 | - | 4 | 1298 | |||
| BC026154.2 | - | 13 | 1654 | 4839268 | ||