Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MCTP1 SIRNA Assays ›
Gene Symbol:
MCTP1
Gene Name:
multiple C2 and transmembrane domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 94703690 - 95284575 on Build GRCh38
UniGene ID:
Hs.591248
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36296
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MCTP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002796.3
NP_001002796.1
14
1820
NM_001297777.1
NP_001284706.1
12
1682
NM_024717.5
NP_078993.4
14
2099
XM_005272082.4
XP_005272139.1
15
2226
XM_005272090.4
XP_005272147.1
12
1461
XM_006714692.3
XP_006714755.1
12
2049
XM_011543649.2
XP_011541951.1
15
2300
XM_011543650.2
XP_011541952.1
13
1277
XM_017009855.1
XP_016865344.1
13
1500
XM_017009856.1
XP_016865345.1
14
1599
XM_017009857.1
XP_016865346.1
13
1346
XM_017009858.1
XP_016865347.1
12
1422
XM_017009859.1
XP_016865348.1
14
1424
XM_017009860.1
XP_016865349.1
13
1521
XM_017009861.1
XP_016865350.1
12
1796
XM_017009862.1
XP_016865351.1
11
1794
XM_017009863.1
XP_016865352.1
11
1422
XM_017009864.1
XP_016865353.1
13
2162
XM_017009865.1
XP_016865354.1
12
1742
GenBank mRNA
AK025997.1
-
3
187
AK057694.1
-
12
1461
AK058012.1
-
13
1366
AK091330.1
-
7
823
AK297325.1
-
11
1102
AY656715.1
-
14
2099
AY656716.1
-
14
1436
BC030005.2
-
14
1581
3834212
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
calcium-mediated signaling
calcium ion binding
calcium-dependent phospholipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline