Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EHMT1 SIRNA Assays ›
Gene Symbol:
EHMT1
Gene Name:
euchromatic histone lysine methyltransferase 1
Gene Aliases:
EUHMTASE1, Eu-HMTase1, FP13812, GLP, GLP1, KMT1D, bA188C12.1
Chromosome Location:
Chr.9: 137618988 - 137836127 on Build GRCh38
UniGene ID:
Hs.495511
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36391
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EHMT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145527.1
NP_001138999.1
12
1872
NM_024757.4
NP_079033.4
12
1872
XM_005266105.4
XP_005266162.1
12
1915
XM_005266110.1
XP_005266167.1
11
1787
XM_006717288.2
XP_006717351.1
12
1833
XM_011519021.2
XP_011517323.1
12
2870
XM_011519022.2
XP_011517324.1
12
2325
XM_011519023.2
XP_011517325.1
12
2849
XM_011519024.2
XP_011517326.1
11
2793
XM_011519025.1
XP_011517327.1
12
2035
XM_011519026.2
XP_011517328.1
11
2728
XM_011519027.2
XP_011517329.1
12
2871
XM_011519028.2
XP_011517330.1
12
2871
XM_011519029.2
XP_011517331.1
4
460
XM_011519033.2
XP_011517335.1
11
2707
XM_017015134.1
XP_016870623.1
12
2303
XM_017015135.1
XP_016870624.1
12
1855
XM_017015136.1
XP_016870625.1
11
1777
XM_017015137.1
XP_016870626.1
12
2014
XM_017015138.1
XP_016870627.1
11
1766
GenBank mRNA
AB028932.1
-
7
692
AB058779.2
-
7
692
AK022941.1
-
11
1787
AY083210.1
-
10
1742
BC011608.2
-
2
89
BC047504.1
-
12
1872
5298029
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
DNA methylation
embryo development
chromatin modification
histone methylation
peptidyl-lysine monomethylation
peptidyl-lysine dimethylation
negative regulation of transcription, DNA-templated
histone H3-K9 methylation
response to fungicide
histone H3-K27 methylation
regulation of signal transduction by p53 class mediator
p53 binding
protein binding
methyltransferase activity
zinc ion binding
protein-lysine N-methyltransferase activity
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K9 specific)
histone methyltransferase activity (H3-K27 specific)
C2H2 zinc finger domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0