Search
Search

MORN1| Gene Symbol: MORN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301060.1 | NP_001287989.1 | 2 | 338 | ||
| NM_024848.2 | NP_079124.1 | 2 | 338 | |||
| XM_011542169.1 | XP_011540471.1 | 2 | 338 | |||
| XM_011542170.2 | XP_011540472.1 | 2 | 338 | |||
| XM_011542173.2 | XP_011540475.1 | 2 | 338 | |||
| XM_011542174.2 | XP_011540476.1 | 2 | 338 | |||
| XM_011542176.1 | XP_011540478.1 | 2 | 338 | |||
| XM_011542177.1 | XP_011540479.1 | 2 | 338 | |||
| XM_017002365.1 | XP_016857854.1 | 2 | 338 | |||
| GenBank mRNA | AK024003.1 | - | 2 | 141 | ||
| BC021704.2 | - | 2 | 294 | 4825983 | ||
| DB041436.1 | - | 2 | 338 | |||