Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRMT2B SIRNA Assays ›
Gene Symbol:
TRMT2B
Gene Name:
tRNA methyltransferase 2 homolog B
Gene Aliases:
CXorf34, dJ341D10.3
Chromosome Location:
Chr.X: 100973358 - 101052118 on Build GRCh38
UniGene ID:
Hs.496501
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36758
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRMT2B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001167970.1
NP_001161442.1
7
979
NM_001167971.1
NP_001161443.1
7
960
NM_001167972.1
NP_001161444.1
6
922
NM_024917.5
NP_079193.2
7
1372
XM_005262195.2
XP_005262252.1
8
1095
XM_005262196.2
XP_005262253.1
6
890
XM_005262201.2
XP_005262258.1
4
470
XM_006724705.2
XP_006724768.1
7
1034
XM_011531042.2
XP_011529344.1
4
588
XM_011531044.2
XP_011529346.1
6
649
XM_017029858.1
XP_016885347.1
7
1367
XM_017029859.1
XP_016885348.1
6
682
XM_017029860.1
XP_016885349.1
7
981
XM_017029861.1
XP_016885350.1
6
891
XM_017029862.1
XP_016885351.1
6
1232
XM_017029863.1
XP_016885352.1
8
904
XM_017029864.1
XP_016885353.1
6
1096
XM_017029865.1
XP_016885354.1
6
724
XM_017029866.1
XP_016885355.1
7
787
XM_017029867.1
XP_016885356.1
7
824
XM_017029868.1
XP_016885357.1
6
697
XM_017029869.1
XP_016885358.1
5
1041
XM_017029870.1
XP_016885359.1
5
631
XM_017029871.1
XP_016885360.1
5
613
XM_017029872.1
XP_016885361.1
6
724
XM_017029873.1
XP_016885362.1
6
699
XM_017029874.1
XP_016885363.1
5
1041
XM_017029875.1
XP_016885364.1
4
588
GenBank mRNA
AK022749.1
-
6
735
AK315780.1
-
7
1129
AL832849.1
-
7
898
BC007526.1
-
6
878
2988022
BC009437.1
-
6
865
3504468
BC020116.1
-
7
933
4342798
BC034272.2
-
7
1337
4933351
BX647969.1
-
6
919
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA methyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
RNA methylation
tRNA processing
protein binding
S-adenosylmethionine-dependent tRNA (m5U54) methyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0