Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEMA6D SIRNA Assays ›
Gene Symbol:
SEMA6D
Gene Name:
semaphorin 6D
Gene Aliases:
-
Chromosome Location:
Chr.15: 47184101 - 47774223 on Build GRCh38
UniGene ID:
Hs.511265
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36853
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEMA6D
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198999.1
NP_001185928.1
14
1483
NM_020858.1
NP_065909.1
11
1471
NM_024966.2
NP_079242.2
11
1471
NM_153616.1
NP_705869.1
11
1471
NM_153617.1
NP_705870.1
11
1471
NM_153618.1
NP_705871.1
11
1471
NM_153619.1
NP_705872.1
11
1471
XM_005254685.3
XP_005254742.1
11
1433
XM_005254686.2
XP_005254743.1
11
1377
XM_005254687.2
XP_005254744.1
11
1377
XM_005254689.3
XP_005254746.1
11
1433
XM_011522075.2
XP_011520377.1
14
1588
XM_011522076.2
XP_011520378.1
11
1377
XM_011522077.2
XP_011520379.1
11
1377
XM_011522078.2
XP_011520380.1
11
1377
XM_011522079.2
XP_011520381.1
11
1377
XM_011522080.2
XP_011520382.1
11
1377
XM_011522081.2
XP_011520383.1
11
1377
XM_017022617.1
XP_016878106.1
14
1401
XM_017022618.1
XP_016878107.1
11
1433
XM_017022619.1
XP_016878108.1
11
1433
XM_017022620.1
XP_016878109.1
14
1588
XM_017022621.1
XP_016878110.1
14
1588
GenBank mRNA
AB040912.2
-
14
1483
AF389426.1
-
11
1471
AF389427.1
-
11
1471
AF389428.1
-
11
1471
AF389429.1
-
11
1471
AF389430.1
-
11
1471
AK021660.1
-
8
769
AK290032.1
-
11
1392
BC150253.1
-
14
1483
9007179
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane-bound signaling molecule
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
neural crest cell migration
positive regulation of smooth muscle cell migration
negative regulation of smooth muscle cell migration
ventricular system development
positive regulation of cell migration
negative regulation of axon extension involved in axon guidance
negative chemotaxis
semaphorin-plexin signaling pathway
semaphorin receptor binding
chemorepellent activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0