Search
Search

ADAMTS20| Gene Symbol: ADAMTS20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025003.3 | NP_079279.3 | 17 | 2402 | ||
| XM_011538754.2 | XP_011537056.1 | 17 | 2455 | |||
| XM_017019979.1 | XP_016875468.1 | 10 | 1471 | |||
| GenBank mRNA | AF488804.1 | - | 17 | 2402 | ||
| AJ515153.3 | - | 17 | 2396 | |||
| AJ515154.2 | - | 17 | 2396 | |||