Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CTNND1 SIRNA Assays ›
Gene Symbol:
CTNND1
TMX2-CTNND1
Gene Name:
catenin delta 1
TMX2-CTNND1 readthrough (NMD candidate)
Gene Aliases:
CAS, CTNND, P120CAS, P120CTN, p120, p120(CAS), p120(CTN)
-
Chromosome Location:
Chr.11: 57761762 - 57819180 on Build GRCh38
Chromosome Location:
Chr.11: 57712523 - 57819180 on Build GRCh38
UniGene ID:
Hs.166011
Hs.166011
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s3725
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CTNND1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001085458.1
NP_001078927.1
14
2800
NM_001085459.1
NP_001078928.1
13
2782
NM_001085460.1
NP_001078929.1
13
2709
NM_001085461.1
NP_001078930.1
13
2782
NM_001085462.1
NP_001078931.1
12
2590
NM_001085463.1
NP_001078932.1
12
2319
NM_001085464.1
NP_001078933.1
11
2301
NM_001085465.1
NP_001078934.1
11
2301
NM_001085466.1
NP_001078935.1
12
2319
NM_001085467.1
NP_001078936.1
11
2374
NM_001085468.1
NP_001078937.1
11
2301
NM_001085469.1
NP_001078938.1
10
2229
NM_001206883.1
NP_001193812.1
13
2499
NM_001206884.1
NP_001193813.1
13
2499
NM_001206885.1
NP_001193814.1
14
2800
NM_001206886.1
NP_001193815.1
12
2481
NM_001206887.1
NP_001193816.1
12
2481
NM_001206888.1
NP_001193817.1
12
2481
NM_001206889.1
NP_001193818.1
13
2600
NM_001206890.1
NP_001193819.1
11
2323
NM_001206891.1
NP_001193820.1
13
2673
NM_001331.2
NP_001322.1
13
2782
GenBank mRNA
AF062317.1
-
13
2747
AF062318.1
-
9
1523
AF062319.1
-
13
2464
AF062320.1
-
13
2464
AF062321.1
-
14
2765
AF062322.1
-
12
2446
AF062323.1
-
12
2284
AF062324.1
-
13
2674
AF062325.1
-
12
2446
AF062326.1
-
8
1505
AF062327.1
-
12
2446
AF062328.1
-
13
2747
AF062329.1
-
13
2565
AF062330.1
-
11
2288
AF062331.1
-
13
2638
AF062332.1
-
11
2266
AF062333.1
-
8
1505
AF062334.1
-
11
2339
AF062335.1
-
11
2266
AF062336.1
-
10
2194
AF062337.1
-
12
2284
AF062338.1
-
11
2266
AF062339.1
-
13
2747
AF062340.1
-
8
1578
AF062341.1
-
14
2765
AF062342.1
-
9
1523
AF062343.1
-
12
2555
AF062344.1
-
8
1505
AK292554.1
-
13
2724
BC010501.1
-
9
1903
BC075795.1
-
13
2598
5269008
BT007333.1
-
7
1242
CR749275.1
-
14
2560
Gene Symbol: TMX2-CTNND1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_037646.1
15
2788
GenBank mRNA
AB002382.1
-
15
2694
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
cell adhesion
brain development
Wnt signaling pathway
single organismal cell-cell adhesion
adherens junction organization
negative regulation of canonical Wnt signaling pathway
receptor binding
protein binding
protein kinase binding
cadherin binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0