Search
Search

FCRL5| Gene Symbol: FCRL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195388.1 | NP_001182317.1 | 4 | 619 | ||
| NM_031281.2 | NP_112571.2 | 4 | 619 | |||
| XM_011510030.1 | XP_011508332.1 | 3 | 321 | |||
| XM_011510031.1 | XP_011508333.1 | 4 | 617 | |||
| XM_011510032.1 | XP_011508334.1 | 4 | 617 | |||
| XM_011510033.1 | XP_011508335.1 | 4 | 617 | |||
| GenBank mRNA | AF343662.1 | - | 4 | 554 | ||
| AF343663.1 | - | 4 | 554 | |||
| AF343664.1 | - | 4 | 554 | |||
| AF369794.2 | - | 4 | 619 | |||
| AF397453.1 | - | 4 | 461 | |||
| AK300010.1 | - | 3 | 345 | |||
| AK301356.1 | - | 5 | 659 | |||
| AL834187.1 | - | 5 | 659 | |||
| BC101066.2 | - | 4 | 489 | 40008649 | ||
| BC101067.1 | - | 4 | 489 | 40008653 | ||
| BC101069.2 | - | 4 | 489 | 40008658 | ||