Search
Search

RBP5| Gene Symbol: RBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017020002.1 | XP_016875491.1 | 2 | 671 | ||
| GenBank mRNA | AF212239.1 | - | 2 | 283 | ||
| AK096947.1 | - | 1 | 582 | |||
| AK223378.1 | - | 2 | 298 | |||
| AK311977.1 | - | 2 | 296 | |||
| AY007436.1 | - | 2 | 279 | |||
| BC029355.1 | - | 2 | 300 | 4613026 | ||