Search
Search

B3GNT5| Gene Symbol: B3GNT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032047.4 | NP_114436.1 | 2 | 684 | ||
| XM_005247823.4 | XP_005247880.1 | 2 | 889 | |||
| XM_005247824.3 | XP_005247881.1 | 3 | 943 | |||
| XM_005247825.3 | XP_005247882.1 | 3 | 827 | |||
| XM_011513225.2 | XP_011511527.1 | 2 | 1323 | |||
| XM_011513226.2 | XP_011511528.1 | 3 | 800 | |||
| XM_011513227.2 | XP_011511529.1 | 3 | 834 | |||
| XM_011513228.2 | XP_011511530.1 | 2 | 735 | |||
| XM_011513229.2 | XP_011511531.1 | 2 | 589 | |||
| XM_017007309.1 | XP_016862798.1 | 3 | 1957 | |||
| GenBank mRNA | AB045278.1 | - | 1 | 283 | ||
| AB209517.1 | - | 2 | 502 | |||
| AF368169.1 | - | 1 | 614 | |||
| AJ304505.1 | - | 1 | 197 | |||
| AK074424.1 | - | 2 | 671 | |||
| BC028058.1 | - | 2 | 677 | 5175670 | ||