Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DCN SIRNA Assays ›
          Gene Symbol:
          DCN
          Gene Name:
          decorin
          Gene Aliases:
          CSCD, DSPG2, PG40, PGII, PGS2, SLRR1B
          Chromosome Location:
          Chr.12: 91143277 - 91183124 on Build GRCh38
          UniGene ID:
          Hs.156316
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s3966

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DCN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001920.4 NP_001911.1 8 1663
          NM_133503.3 NP_598010.1 8 1450
          NM_133504.3 NP_598011.1 5 927
          NM_133505.3 NP_598012.1 4 813
          NM_133506.3 NP_598013.1 3 693
          NM_133507.3 NP_598014.1 2 580
          XM_005268693.1 XP_005268750.1 8 1416
          XM_006719270.1 XP_006719333.1 8 1239
          XM_017018917.1 XP_016874406.1 8 1390
          GenBank mRNA AK291309.1 - 8 1322
          BC005322.1 - 7 1201 3934022
          BX571756.1 - 8 1377

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          placenta development
          negative regulation of protein kinase activity
          skeletal muscle tissue development
          aging
          response to mechanical stimulus
          organ morphogenesis
          positive regulation of autophagy
          negative regulation of endothelial cell migration
          positive regulation of phosphatidylinositol 3-kinase signaling
          positive regulation of macroautophagy
          negative regulation of angiogenesis
          cytokine-mediated signaling pathway
          peptide cross-linking via chondroitin 4-sulfate glycosaminoglycan
          extracellular matrix disassembly
          extracellular matrix organization
          glycosaminoglycan metabolic process
          chondroitin sulfate biosynthetic process
          chondroitin sulfate catabolic process
          dermatan sulfate biosynthetic process
          response to lipopolysaccharide
          wound healing
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of JAK-STAT cascade
          positive regulation of mitochondrial depolarization
          positive regulation of mitochondrial fission
          negative regulation of vascular endothelial growth factor signaling pathway
          protein kinase inhibitor activity
          protein binding
          collagen binding
          glycosaminoglycan binding
          poly(A) RNA binding
          protein N-terminus binding
          extracellular matrix binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0