Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACE SIRNA Assays ›
          Gene Symbol:
          ACE
          Gene Name:
          angiotensin I converting enzyme
          Gene Aliases:
          ACE1, CD143, DCP, DCP1, ICH, MVCD3
          Chromosome Location:
          Chr.17: 63477061 - 63498380 on Build GRCh38
          UniGene ID:
          Hs.298469
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s3967

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACE
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000789.3 NP_000780.1 16 2450
          NM_001178057.1 NP_001171528.1 5 745
          NM_152830.2 NP_690043.1 5 745
          XM_006721737.3 XP_006721800.2 5 858
          GenBank mRNA AB208971.1 - 16 2440
          AK301988.1 - 5 716
          AK302019.1 - 5 619
          BM908222.1 - 5 757
          J04144.1 - 16 2438
          JQ927553.1 - 16 2450
          M26657.1 - 5 716
          M29981.1 - 5 794
          X16295.1 - 5 722

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          blood vessel remodeling
          angiotensin maturation
          angiotensin catabolic process in blood
          regulation of renal output by angiotensin
          neutrophil mediated immunity
          antigen processing and presentation of peptide antigen via MHC class I
          regulation of systemic arterial blood pressure by renin-angiotensin
          spermatogenesis
          regulation of blood pressure
          regulation of smooth muscle cell migration
          regulation of vasoconstriction
          mononuclear cell proliferation
          regulation of vasodilation
          hormone catabolic process
          peptide catabolic process
          beta-amyloid metabolic process
          arachidonic acid secretion
          heart contraction
          regulation of angiotensin metabolic process
          hematopoietic stem cell differentiation
          positive regulation of protein tyrosine kinase activity
          cell proliferation in bone marrow
          positive regulation of peptidyl-tyrosine autophosphorylation
          regulation of hematopoietic stem cell proliferation
          negative regulation of gap junction assembly
          positive regulation of peptidyl-cysteine S-nitrosylation
          endopeptidase activity
          carboxypeptidase activity
          drug binding
          metallopeptidase activity
          exopeptidase activity
          tripeptidyl-peptidase activity
          peptidyl-dipeptidase activity
          zinc ion binding
          chloride ion binding
          mitogen-activated protein kinase kinase binding
          bradykinin receptor binding
          mitogen-activated protein kinase binding
          metallodipeptidase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0