Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACBD5 SIRNA Assays ›
          Gene Symbol:
          ACBD5
          Gene Name:
          acyl-CoA binding domain containing 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 27182624 - 27248674 on Build GRCh38
          UniGene ID:
          Hs.530597
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s40668

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACBD5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042473.3 NP_001035938.1 7 785
          NM_001271512.3 NP_001258441.1 6 1099
          NM_001301251.1 NP_001288180.1 6 664
          NM_001301252.1 NP_001288181.1 5 978
          NM_001301253.1 NP_001288182.1 6 654
          NM_145698.4 NP_663736.2 7 910
          XM_006717528.3 XP_006717591.3 8 7343
          XM_006717530.3 XP_006717593.1 8 942
          XM_006717531.2 XP_006717594.1 8 952
          XM_006717535.2 XP_006717598.1 8 809
          XM_017016884.1 XP_016872373.1 8 7343
          XM_017016885.1 XP_016872374.1 7 7310
          XM_017016886.1 XP_016872375.1 7 7310
          XM_017016887.1 XP_016872376.1 8 873
          XM_017016888.1 XP_016872377.1 8 938
          XM_017016889.1 XP_016872378.1 7 840
          XM_017016891.1 XP_016872380.1 7 905
          XM_017016892.1 XP_016872381.1 7 826
          XM_017016895.1 XP_016872384.1 8 809
          XM_017016896.1 XP_016872385.1 7 777
          XM_017016900.1 XP_016872389.1 7 727
          XM_017016901.1 XP_016872390.1 7 707
          XM_017016902.1 XP_016872391.1 7 726
          XM_017016903.1 XP_016872392.1 7 711
          XM_017016904.1 XP_016872393.1 6 706
          XM_017016905.1 XP_016872394.1 6 692
          XM_017016907.1 XP_016872396.1 6 706
          XM_017016908.1 XP_016872397.1 6 695
          GenBank mRNA AB082527.1 - 6 613
          AF505653.1 - 6 651
          AK057469.1 - 7 777
          AK295031.1 - 6 736
          AK304842.1 - 5 629
          BC030555.1 - 7 814 5218685

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          macroautophagy
          pexophagy
          aggrephagy
          fatty-acyl-CoA binding
          molecular_function
          lipid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0