Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYOCD SIRNA Assays ›
          Gene Symbol:
          MYOCD
          Gene Name:
          myocardin
          Gene Aliases:
          MYCD
          Chromosome Location:
          Chr.17: 12665890 - 12768949 on Build GRCh38
          UniGene ID:
          Hs.462257
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s41184

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MYOCD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001146312.2 NP_001139784.1 14 2867
          NM_153604.3 NP_705832.1 13 2723
          XM_005256863.1 XP_005256920.1 13 2738
          XM_005256864.1 XP_005256921.1 15 2911
          XM_017025342.1 XP_016880831.1 13 2741
          GenBank mRNA AF532596.1 - 13 2424
          AK095846.1 - 1 29
          AK128608.1 - 14 1833
          AK292885.1 - 14 2736
          AY764180.1 - 14 2860
          BC126307.1 - 14 2626 8992023
          BC143391.1 - 14 2626 9051897
          BX640673.1 - 6 1554

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of cell growth by extracellular stimulus
          vasculogenesis
          response to hypoxia
          cardiac ventricle development
          positive regulation of transcription from RNA polymerase II promoter involved in myocardial precursor cell differentiation
          transcription from RNA polymerase II promoter
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          negative regulation of cardiac muscle cell apoptotic process
          negative regulation of myotube differentiation
          positive regulation of transforming growth factor beta receptor signaling pathway
          cardiocyte differentiation
          regulation of histone acetylation
          hepatic stellate cell activation
          vascular smooth muscle cell differentiation
          positive regulation of DNA binding
          cellular component maintenance
          regulation of myoblast differentiation
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of smooth muscle contraction
          lung alveolus development
          digestive tract development
          positive regulation of sequence-specific DNA binding transcription factor activity
          smooth muscle cell differentiation
          regulation of smooth muscle cell differentiation
          positive regulation of smooth muscle cell differentiation
          cardiac muscle cell differentiation
          ventricular cardiac muscle cell differentiation
          uterus development
          urinary bladder development
          cell growth involved in cardiac muscle cell development
          cellular response to growth factor stimulus
          cellular response to hypoxia
          ductus arteriosus closure
          negative regulation of beta-amyloid clearance
          regulation of phenotypic switching
          positive regulation of transcription from RNA polymerase II promoter involved in smooth muscle cell differentiation
          positive regulation of cardiac vascular smooth muscle cell differentiation
          positive regulation of cardiac muscle cell differentiation
          negative regulation of skeletal muscle cell differentiation
          core promoter sequence-specific DNA binding
          transcription factor activity, RNA polymerase II transcription factor binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription coactivator activity
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          protein binding
          transcription factor binding
          histone acetyltransferase binding
          histone deacetylase binding
          R-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0