Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AZIN2 SIRNA Assays ›
Gene Symbol:
AZIN2
Gene Name:
antizyme inhibitor 2
Gene Aliases:
ADC, AZI2, AZIB1, ODC-p, ODC1L, ODCp
Chromosome Location:
Chr.1: 33081110 - 33162288 on Build GRCh38
UniGene ID:
Hs.101807
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s41478
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: AZIN2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001293562.1
NP_001280491.1
5
664
NM_001301823.1
NP_001288752.1
3
567
NM_001301824.1
NP_001288753.1
2
393
NM_001301825.1
NP_001288754.1
3
364
NM_001301826.1
NP_001288755.1
3
364
NM_052998.3
NP_443724.1
6
879
NR_126031.1
3
364
XM_005270404.2
XP_005270461.1
5
626
XM_005270406.1
XP_005270463.1
4
493
XM_005270407.1
XP_005270464.1
3
263
XM_006710312.1
XP_006710375.1
3
274
XM_011540557.1
XP_011538859.1
4
445
XM_011540563.1
XP_011538865.1
5
624
XM_011540566.1
XP_011538868.1
5
624
XM_017000167.1
XP_016855656.1
5
624
XM_017000168.1
XP_016855657.1
6
749
XM_017000169.1
XP_016855658.1
6
657
XM_017000170.1
XP_016855659.1
5
651
XM_017000171.1
XP_016855660.1
6
737
XM_017000172.1
XP_016855661.1
6
654
XM_017000173.1
XP_016855662.1
4
330
XM_017000174.1
XP_016855663.1
5
624
XM_017000175.1
XP_016855664.1
4
449
XM_017000176.1
XP_016855665.1
4
386
XM_017000177.1
XP_016855666.1
4
444
XM_017000178.1
XP_016855667.1
4
397
GenBank mRNA
AB075825.1
-
2
2482
AK057051.1
-
2
367
AK095127.1
-
3
548
AK122697.1
-
2
1235
AK315677.1
-
5
626
AL832640.2
-
5
1415
AY050634.1
-
3
292
AY050635.1
-
3
292
AY050636.1
-
3
292
AY050637.1
-
3
292
AY325129.1
-
5
610
BC010449.1
-
5
610
4156927
BC028128.1
-
6
879
5164244
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: HQ413774, BC078981.
This siRNA also targets the following transcripts in Mouse: AK077201, AK148717, AK047522, BC138973, BC138974, and others.
Panther Classification:
Molecular Function -
decarboxylase
Gene Ontology Categories:
Function(s)
Process(es)
spermatogenesis
putrescine transport
putrescine biosynthetic process from ornithine
negative regulation of protein catabolic process
positive regulation of catalytic activity
agmatine biosynthetic process
trans-Golgi network membrane organization
positive regulation of polyamine transmembrane transport
protein binding
arginine decarboxylase activity
putrescine transmembrane transporter activity
ornithine decarboxylase activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0