Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFYVE27 SIRNA Assays ›
Gene Symbol:
ZFYVE27
Gene Name:
zinc finger FYVE-type containing 27
Gene Aliases:
PROTRUDIN, SPG33
Chromosome Location:
Chr.10: 97737121 - 97760907 on Build GRCh38
UniGene ID:
Hs.744075
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s42249
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFYVE27
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002261.3
NP_001002261.1
6
700
NM_001002262.3
NP_001002262.1
7
905
NM_001174119.1
NP_001167590.1
6
805
NM_001174120.1
NP_001167591.1
5
643
NM_001174121.1
NP_001167592.1
5
632
NM_001174122.1
NP_001167593.1
4
547
NM_144588.6
NP_653189.3
7
905
XM_005269502.3
XP_005269559.1
7
894
XM_005269503.3
XP_005269560.1
7
898
XM_005269504.3
XP_005269561.1
7
894
XM_005269505.4
XP_005269562.1
7
894
XM_005269506.3
XP_005269563.1
7
894
XM_005269508.3
XP_005269565.1
6
823
XM_005269509.3
XP_005269566.1
6
827
XM_005269510.3
XP_005269567.1
5
639
XM_005269511.3
XP_005269568.1
5
645
XM_011539252.2
XP_011537554.1
7
894
XM_011539253.2
XP_011537555.1
7
894
XM_011539254.2
XP_011537556.1
7
869
XM_011539255.2
XP_011537557.1
7
876
XM_017015644.1
XP_016871133.1
7
894
XM_017015645.1
XP_016871134.1
7
894
XM_017015646.1
XP_016871135.1
7
894
XM_017015647.1
XP_016871136.1
7
898
XM_017015648.1
XP_016871137.1
5
639
XM_017015649.1
XP_016871138.1
5
639
XM_017015650.1
XP_016871139.1
7
869
XM_017015651.1
XP_016871140.1
6
823
XM_017015652.1
XP_016871141.1
6
827
XM_017015653.1
XP_016871142.1
6
823
XM_017015654.1
XP_016871143.1
6
827
XM_017015655.1
XP_016871144.1
5
645
GenBank mRNA
AK057481.1
-
7
876
AK074876.1
-
7
886
AK097945.1
-
5
636
AK296295.1
-
5
632
AK296588.1
-
6
750
AK299735.1
-
4
540
AK300438.1
-
7
898
AL561314.3
-
7
817
AL834235.1
-
6
634
BC030621.2
-
7
898
4818199
BU838354.1
-
3
223
BX647836.1
-
7
840
KU178847.1
-
5
495
KU178848.1
-
5
495
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
neuron projection development
neurotrophin TRK receptor signaling pathway
protein localization to plasma membrane
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0