Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZFYVE27 SIRNA Assays ›
          Gene Symbol:
          ZFYVE27
          Gene Name:
          zinc finger FYVE-type containing 27
          Gene Aliases:
          PROTRUDIN, SPG33
          Chromosome Location:
          Chr.10: 97737121 - 97760907 on Build GRCh38
          UniGene ID:
          Hs.744075
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s42250

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZFYVE27
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001002261.3 NP_001002261.1 10, 11 1116
          NM_001002262.3 NP_001002262.1 10, 11 1285
          NM_001174119.1 NP_001167590.1 9, 10 1185
          NM_001174120.1 NP_001167591.1 8, 9 1023
          NM_001174121.1 NP_001167592.1 8, 9 1027
          NM_001174122.1 NP_001167593.1 7, 8 927
          NM_144588.6 NP_653189.3 11, 12 1306
          XM_005269502.3 XP_005269559.1 11, 12 1310
          XM_005269503.3 XP_005269560.1 11, 12 1314
          XM_005269504.3 XP_005269561.1 11, 12 1295
          XM_005269505.4 XP_005269562.1 10, 11 1289
          XM_005269506.3 XP_005269563.1 10, 11 1274
          XM_005269508.3 XP_005269565.1 10, 11 1239
          XM_005269509.3 XP_005269566.1 10, 11 1243
          XM_005269510.3 XP_005269567.1 9, 10 1055
          XM_005269511.3 XP_005269568.1 9, 10 1061
          XM_011539252.2 XP_011537554.1 11, 12 1310
          XM_011539253.2 XP_011537555.1 10, 11 1289
          XM_011539254.2 XP_011537556.1 11, 12 1285
          XM_011539255.2 XP_011537557.1 11, 12 1292
          XM_017015647.1 XP_016871136.1 10, 11 1293
          XM_017015648.1 XP_016871137.1 9, 10 1040
          XM_017015649.1 XP_016871138.1 8, 9 1034
          XM_017015650.1 XP_016871139.1 11, 12 1270
          XM_017015651.1 XP_016871140.1 10, 11 1224
          XM_017015652.1 XP_016871141.1 10, 11 1228
          XM_017015653.1 XP_016871142.1 9, 10 1203
          XM_017015654.1 XP_016871143.1 9, 10 1207
          XM_017015655.1 XP_016871144.1 9, 10 1046
          GenBank mRNA AK074876.1 - 10, 11 1266
          AK097945.1 - 8, 9 1016
          AK296295.1 - 8, 9 1027
          AK296588.1 - 9, 10 1130
          AK299735.1 - 7, 8 920
          AK300438.1 - 10, 11 1363
          AL834235.1 - 10, 11 1050
          BC030621.2 - 11, 12 1299 4818199
          BM544601.1 - 2, 3 228
          BU838354.1 - 6, 7 603
          BX647836.1 - 9, 10 6718
          KU178847.1 - 9, 10 896
          KU178848.1 - 8, 9 875

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane traffic protein membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron projection development
          neurotrophin TRK receptor signaling pathway
          protein localization to plasma membrane
          protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0