Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HYAL2 SIRNA Assays ›
          Gene Symbol:
          HYAL2
          Gene Name:
          hyaluronoglucosaminidase 2
          Gene Aliases:
          LUCA2
          Chromosome Location:
          Chr.3: 50317790 - 50322850 on Build GRCh38
          UniGene ID:
          Hs.76873
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s429
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HYAL2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003773.4 NP_003764.3 4 1917
          NM_033158.4 NP_149348.2 5 2325
          XM_005265524.2 XP_005265581.1 5 2041
          XM_005265525.2 XP_005265582.1 4 2055
          GenBank mRNA AF070608.1 - 3 1633
          AJ000099.1 - 4 1770
          AK092449.1 - 5 2204
          AK127945.1 - 3 3966
          AK225635.1 - 4 1548
          BC000692.2 - 4 1822 3347760
          U09577.1 - 4 1779

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00186841_m1

          Panther Classification:

          Molecular Function -

          glycosidase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          kidney development
          hematopoietic progenitor cell differentiation
          carbohydrate metabolic process
          glycosaminoglycan catabolic process
          response to virus
          multicellular organism aging
          negative regulation of fibroblast migration
          fusion of virus membrane with host plasma membrane
          transformation of host cell by virus
          hyaluronan catabolic process
          negative regulation of cell growth
          positive regulation of urine volume
          monocyte activation
          positive regulation of protein import into nucleus
          negative regulation of MAP kinase activity
          cellular response to fibroblast growth factor stimulus
          positive regulation of transcription from RNA polymerase II promoter
          response to antibiotic
          viral entry into host cell
          skeletal system morphogenesis
          positive regulation of inflammatory response
          cartilage development
          defense response to virus
          negative regulation of protein kinase B signaling
          multicellular organismal iron ion homeostasis
          negative regulation of protein tyrosine kinase activity
          renal water absorption
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to UV-B
          cellular response to transforming growth factor beta stimulus
          positive regulation of interleukin-8 secretion
          positive regulation of interleukin-6 secretion
          positive regulation of extrinsic apoptotic signaling pathway
          RNA polymerase II transcription coactivator activity
          virus receptor activity
          hyalurononglucosaminidase activity
          protein binding
          hyaluronic acid binding
          enzyme binding
          receptor signaling protein tyrosine kinase inhibitor activity
          receptor tyrosine kinase binding
          hyaluronoglucuronidase activity
          transforming growth factor beta binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0