Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CTNNB1 SIRNA Assays ›
          Gene Symbol:
          CTNNB1
          Gene Name:
          catenin beta 1
          Gene Aliases:
          CTNNB, MRD19, armadillo
          Chromosome Location:
          Chr.3: 41199451 - 41240448 on Build GRCh38
          UniGene ID:
          Hs.476018
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s436
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CTNNB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098209.1 NP_001091679.1 7 1262
          NM_001098210.1 NP_001091680.1 7 1262
          NM_001904.3 NP_001895.1 7 1262
          XM_005264886.2 XP_005264943.1 7 1276
          XM_006712983.1 XP_006713046.1 7 1174
          XM_006712984.1 XP_006713047.1 7 1174
          XM_006712985.1 XP_006713048.1 7 1236
          XM_017005738.1 XP_016861227.1 7 1276
          GenBank mRNA AB062292.1 - 2 225
          AB451264.1 - 5 994
          AB451392.1 - 5 994
          AK289932.1 - 8 1150
          AK294778.1 - 8 1375
          AK296790.1 - 7 1082
          AK299954.1 - 7 944
          AK314776.1 - 7 1208
          BC058926.1 - 7 1204 6151332
          X87838.1 - 7 1208
          Z19054.1 - 7 1194

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00170025_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          embryonic axis specification
          patterning of blood vessels
          branching involved in ureteric bud morphogenesis
          in utero embryonic development
          gastrulation with mouth forming second
          cell fate specification
          endodermal cell fate commitment
          neuron migration
          epithelial to mesenchymal transition
          neural plate development
          liver development
          positive regulation of neuroblast proliferation
          positive regulation of mesenchymal cell proliferation
          lens morphogenesis in camera-type eye
          regulation of secondary heart field cardioblast proliferation
          metanephros morphogenesis
          negative regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis
          transcription, DNA-templated
          cell adhesion
          cell-matrix adhesion
          Wnt signaling pathway, calcium modulating pathway
          chemical synaptic transmission
          ectoderm development
          glial cell fate determination
          midgut development
          negative regulation of cell proliferation
          anterior/posterior axis specification
          dorsal/ventral axis specification
          proximal/distal pattern formation
          positive regulation of epithelial to mesenchymal transition
          positive regulation of heparan sulfate proteoglycan biosynthetic process
          Schwann cell proliferation
          response to activity
          Wnt signaling pathway
          single organismal cell-cell adhesion
          stem cell population maintenance
          layer formation in cerebral cortex
          central nervous system vasculogenesis
          osteoclast differentiation
          androgen receptor signaling pathway
          male genitalia development
          hindbrain development
          regulation of centriole-centriole cohesion
          pancreas development
          hair follicle morphogenesis
          regulation of myelination
          positive regulation of telomere maintenance via telomerase
          negative regulation of chondrocyte differentiation
          response to estradiol
          positive regulation of type I interferon production
          T cell differentiation in thymus
          negative regulation of protein sumoylation
          response to cytokine
          adherens junction assembly
          protein localization to cell surface
          embryonic heart tube development
          genitalia morphogenesis
          embryonic forelimb morphogenesis
          embryonic hindlimb morphogenesis
          hair cell differentiation
          catenin import into nucleus
          embryonic skeletal limb joint morphogenesis
          regulation of T cell proliferation
          odontogenesis of dentin-containing tooth
          response to drug
          embryonic digit morphogenesis
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of MAPK cascade
          positive regulation of neuron apoptotic process
          response to estrogen
          canonical Wnt signaling pathway involved in positive regulation of epithelial to mesenchymal transition
          canonical Wnt signaling pathway involved in negative regulation of apoptotic process
          cellular response to fibroblast growth factor stimulus
          myoblast differentiation
          bone resorption
          positive regulation of endothelial cell differentiation
          positive regulation of osteoblast differentiation
          negative regulation of osteoclast differentiation
          positive regulation of fibroblast growth factor receptor signaling pathway
          regulation of angiogenesis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of mitotic cell cycle, embryonic
          response to cadmium ion
          chromatin-mediated maintenance of transcription
          regulation of fibroblast proliferation
          cell maturation
          synaptic vesicle transport
          thymus development
          oocyte development
          embryonic foregut morphogenesis
          positive regulation of skeletal muscle tissue development
          regulation of smooth muscle cell proliferation
          embryonic cranial skeleton morphogenesis
          negative regulation of oligodendrocyte differentiation
          regulation of neurogenesis
          synapse organization
          positive regulation of sequence-specific DNA binding transcription factor activity
          smooth muscle cell differentiation
          positive regulation of muscle cell differentiation
          protein heterooligomerization
          positive regulation of histone H3-K4 methylation
          positive regulation of telomerase activity
          oviduct development
          canonical Wnt signaling pathway
          trachea formation
          epithelial tube branching involved in lung morphogenesis
          lung cell differentiation
          lung-associated mesenchyme development
          lung induction
          epithelial cell differentiation involved in prostate gland development
          positive regulation of epithelial cell proliferation involved in prostate gland development
          hair follicle placode formation
          mesenchymal cell proliferation involved in lung development
          endothelial tube morphogenesis
          fungiform papilla formation
          canonical Wnt signaling pathway involved in positive regulation of cardiac outflow tract cell proliferation
          sympathetic ganglion development
          cranial ganglion development
          regulation of centromeric sister chromatid cohesion
          cellular response to mechanical stimulus
          cellular response to lithium ion
          cellular response to growth factor stimulus
          cellular response to indole-3-methanol
          renal vesicle formation
          renal inner medulla development
          renal outer medulla development
          nephron tubule formation
          regulation of nephron tubule epithelial cell differentiation
          regulation of calcium ion import
          negative regulation of neuron death
          positive regulation of chromatin-mediated maintenance of transcription
          regulation of euchromatin binding
          regulation of core promoter binding
          beta-catenin-TCF complex assembly
          beta-catenin destruction complex disassembly
          midbrain dopaminergic neuron differentiation
          canonical Wnt signaling pathway involved in midbrain dopaminergic neuron differentiation
          cellular response to insulin-like growth factor stimulus
          embryonic brain development
          dorsal root ganglion development
          regulation of protein localization to cell surface
          positive regulation of determination of dorsal identity
          positive regulation of DNA-templated transcription, initiation
          negative regulation of apoptotic signaling pathway
          RNA polymerase II transcription factor binding
          RNA polymerase II activating transcription factor binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          signal transducer activity
          protein binding
          protein C-terminus binding
          transcription factor binding
          enzyme binding
          kinase binding
          protein kinase binding
          protein phosphatase binding
          estrogen receptor binding
          protein complex binding
          ionotropic glutamate receptor binding
          nuclear hormone receptor binding
          transcription regulatory region DNA binding
          ion channel binding
          alpha-catenin binding
          cadherin binding
          SMAD binding
          androgen receptor binding
          nitric-oxide synthase binding
          I-SMAD binding
          repressing transcription factor binding
          cadherin binding involved in cell-cell adhesion
          euchromatin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0