Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DVL1 SIRNA Assays ›
          Gene Symbol:
          DVL1
          Gene Name:
          dishevelled segment polarity protein 1
          Gene Aliases:
          DRS2, DVL, DVL1L1, DVL1P1
          Chromosome Location:
          Chr.1: 1335278 - 1349142 on Build GRCh38
          UniGene ID:
          Hs.731450
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s4393

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DVL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004421.2 NP_004412.2 14 1642
          XM_005244731.3 XP_005244788.1 14 1747
          XM_005244732.3 XP_005244789.1 14 1747
          XM_005244733.3 XP_005244790.1 14 1671
          GenBank mRNA AB209210.1 - 9 1665
          AF006011.1 - 14 1598
          AK093189.1 - 7 882
          AK095867.1 - 7 860
          AK293286.1 - 14 1448
          AK300063.1 - 6 644
          BC017225.2 - 12 1367
          BC050454.1 - 8 987 6201003
          U46461.1 - 14 1739

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of neurotransmitter levels
          positive regulation of protein phosphorylation
          transcription from RNA polymerase II promoter
          negative regulation of protein kinase activity
          neurotransmitter secretion
          axon guidance
          synapse assembly
          heart development
          neuromuscular junction development
          positive regulation of neuron projection development
          neural tube development
          convergent extension involved in neural plate elongation
          positive regulation of Wnt signaling pathway
          cytoplasmic microtubule organization
          negative regulation of protein binding
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          protein localization to nucleus
          social behavior
          protein localization to microtubule
          intracellular signal transduction
          receptor clustering
          positive regulation of transcription, DNA-templated
          collateral sprouting
          axon extension
          dendrite morphogenesis
          synapse organization
          protein stabilization
          canonical Wnt signaling pathway
          Wnt signaling pathway, planar cell polarity pathway
          prepulse inhibition
          skeletal muscle acetylcholine-gated channel clustering
          negative regulation of canonical Wnt signaling pathway
          cochlea morphogenesis
          planar cell polarity pathway involved in neural tube closure
          positive regulation of canonical Wnt signaling pathway
          positive regulation of protein localization to synapse
          regulation of cellular protein localization
          beta-catenin destruction complex disassembly
          molecular_function
          frizzled binding
          protein binding
          beta-catenin binding
          enzyme binding
          protein kinase binding
          identical protein binding
          Rac GTPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0