Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EDN3 SIRNA Assays ›
          Gene Symbol:
          EDN3
          Gene Name:
          endothelin 3
          Gene Aliases:
          ET-3, ET3, HSCR4, PPET3, WS4B
          Chromosome Location:
          Chr.20: 59300415 - 59325992 on Build GRCh38
          UniGene ID:
          Hs.1408
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s4463

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EDN3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001302455.1 NP_001289384.1 2, 3 757
          NM_001302456.1 NP_001289385.1 2, 3 757
          NM_207032.2 NP_996915.1 2, 3 757
          NM_207033.2 NP_996916.1 2, 3 757
          NM_207034.2 NP_996917.1 2, 3 757
          XM_005260312.4 XP_005260369.1 2, 3 767
          XM_005260313.4 XP_005260370.1 2, 3 769
          XM_006723734.3 XP_006723797.1 2, 3 768
          XM_011528655.2 XP_011526957.1 2, 3 768
          GenBank mRNA BC008876.2 - 2, 3 757 4139312
          BC053866.1 - 2, 3 741 5202210
          BT007085.1 - 2, 3 371
          DN989853.1 - 2, 3 420
          DQ096281.1 - 2, 3 371
          J05081.1 - 2, 3 564
          S50864.1 - 20
          S50866.1 - 20
          X52001.1 - 2, 3 481

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          neural crest cell migration
          positive regulation of leukocyte chemotaxis
          regulation of systemic arterial blood pressure by endothelin
          cellular calcium ion homeostasis
          signal transduction
          cell surface receptor signaling pathway
          cell-cell signaling
          multicellular organism development
          blood circulation
          positive regulation of cell proliferation
          positive regulation of heart rate
          regulation of gene expression
          cellular magnesium ion homeostasis
          artery smooth muscle contraction
          vein smooth muscle contraction
          regulation of vasoconstriction
          peptide hormone secretion
          neuron differentiation
          melanocyte differentiation
          neutrophil chemotaxis
          vasoconstriction
          positive regulation of MAP kinase activity
          positive regulation of cell differentiation
          positive regulation of mitotic nuclear division
          positive regulation of hormone secretion
          inositol phosphate-mediated signaling
          regulation of developmental pigmentation
          positive regulation of potassium ion transmembrane transport
          receptor binding
          hormone activity
          endothelin B receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0