Search
Search

GLT1D1| Gene Symbol: GLT1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144669.2 | NP_653270.1 | 2 | 284 | ||
| NR_133646.1 | 2 | 284 | ||||
| XM_005253553.4 | XP_005253610.2 | 2 | 395 | |||
| XM_011537956.2 | XP_011536258.2 | 2 | 395 | |||
| XM_017018855.1 | XP_016874344.1 | 2 | 395 | |||
| XM_017018857.1 | XP_016874346.1 | 2 | 151 | |||
| XM_017018858.1 | XP_016874347.1 | 2 | 151 | |||
| XM_017018859.1 | XP_016874348.1 | 2 | 395 | |||
| XM_017018860.1 | XP_016874349.1 | 2 | 151 | |||
| XM_017018861.1 | XP_016874350.1 | 2 | 183 | |||
| GenBank mRNA | AK056540.1 | - | 2 | 182 | ||
| AK295333.1 | - | 2 | 112 | |||
| BC043528.1 | - | 2 | 284 | 5168267 | ||