Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RC3H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300850.1 | NP_001287779.1 | 5 | 979 | ||
NM_001300851.1 | NP_001287780.1 | 5 | 979 | |||
NM_001300852.1 | NP_001287781.1 | 5 | 979 | |||
NM_172071.3 | NP_742068.1 | 5 | 979 | |||
XM_005244921.3 | XP_005244978.1 | 5 | 979 | |||
XM_011509231.2 | XP_011507533.1 | 5 | 979 | |||
GenBank mRNA | AB095945.1 | - | 5 | 820 | ||
AK122948.1 | - | 5 | 979 | |||
BC136784.1 | - | 5 | 933 | 9020774 | ||
BC144408.1 | - | 5 | 933 | 9052929 |