Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC24 SIRNA Assays ›
Gene Symbol:
CCDC24
Gene Name:
coiled-coil domain containing 24
Gene Aliases:
-
Chromosome Location:
Chr.1: 43991247 - 43996528 on Build GRCh38
UniGene ID:
Hs.632394
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s45305
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC24
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_152499.2
NP_689712.1
6
712
XM_017000423.1
XP_016855912.1
5
926
XM_017000424.1
XP_016855913.1
5
1063
XM_017000425.1
XP_016855914.1
5
926
XM_017000426.1
XP_016855915.1
5
833
XM_017000427.1
XP_016855916.1
6
830
XM_017000428.1
XP_016855917.1
6
827
XM_017000429.1
XP_016855918.1
5
818
XM_017000430.1
XP_016855919.1
6
827
XM_017000431.1
XP_016855920.1
6
719
XM_017000432.1
XP_016855921.1
5
902
XM_017000433.1
XP_016855922.1
6
683
XM_017000434.1
XP_016855923.1
6
683
XM_017000435.1
XP_016855924.1
5
671
XM_017000436.1
XP_016855925.1
5
671
XM_017000437.1
XP_016855926.1
6
629
XM_017000438.1
XP_016855927.1
6
614
XM_017000439.1
XP_016855928.1
6
590
XM_017000440.1
XP_016855929.1
6
575
XM_017000441.1
XP_016855930.1
6
575
XM_017000442.1
XP_016855931.1
5
455
XM_017000445.1
XP_016855934.1
4
327
XM_017000446.1
XP_016855935.1
4
405
GenBank mRNA
AK024289.1
-
1
1218
AK092569.1
-
5
1912
AY487420.1
-
5
433
BC033868.1
-
6
712
5207706
BC047313.1
-
5
909
BX641142.1
-
4
377
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0