Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MEI1 SIRNA Assays ›
Gene Symbol:
MEI1
Gene Name:
meiotic double-stranded break formation protein 1
Gene Aliases:
SPATA38
Chromosome Location:
Chr.22: 41699503 - 41799456 on Build GRCh38
UniGene ID:
Hs.116419
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s45425
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MEI1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_152513.3
NP_689726.3
21
2608
XM_011529935.1
XP_011528237.1
22
2581
XM_011529936.1
XP_011528238.1
21
2617
XM_011529937.1
XP_011528239.1
21
2617
XM_011529938.1
XP_011528240.1
20
2500
XM_011529939.1
XP_011528241.1
21
2617
XM_011529940.1
XP_011528242.1
21
2617
XM_011529941.1
XP_011528243.1
21
2617
XM_011529942.2
XP_011528244.1
21
2808
XM_011529943.1
XP_011528245.1
20
2341
XM_011529944.1
XP_011528246.1
21
2617
XM_011529945.2
XP_011528247.1
21
2619
XM_011529946.1
XP_011528248.1
18
2185
XM_011529947.1
XP_011528249.1
16
2095
XM_011529948.2
XP_011528250.1
17
1989
XM_011529949.1
XP_011528251.1
16
1926
XM_011529950.1
XP_011528252.1
21
2617
XM_011529952.1
XP_011528254.1
16
2063
XM_011529954.1
XP_011528256.1
11
1367
XM_017028633.1
XP_016884122.1
21
2617
XM_017028635.1
XP_016884124.1
7
947
GenBank mRNA
AK092934.1
-
7
977
AK093341.1
-
1
94
AK094461.1
-
3
394
AK097834.1
-
1
94
AK128158.1
-
5
5313
AK301435.1
-
6
814
AK303957.1
-
5
780
AY952376.1
-
6
1320
AY952377.1
-
6
1320
BC029582.1
-
2
357
BC070111.1
-
17
2193
Back To Top
More Information
Gene Ontology Categories:
Function(s)
meiosis I
male meiosis I
spermatid development
meiotic telomere clustering
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0