Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EIF5A SIRNA Assays ›
          Gene Symbol:
          EIF5A EIF5AL1
          Gene Name:
          eukaryotic translation initiation factor 5A
          eukaryotic translation initiation factor 5A-like 1
          Gene Aliases:
          EIF-5A, EIF5A1, eIF5AI
          EIF5AP1, bA342M3.3
          Chromosome Location:
          Chr.17: 7306999 - 7312463 on Build GRCh38
          Chromosome Location:
          Chr.10: 79512601 - 79516440 on Build GRCh38
          UniGene ID:
          Hs.534314
          Hs.104825
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s4596

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EIF5A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001143760.1 NP_001137232.1 2 208
          NM_001143761.1 NP_001137233.1 2 236
          NM_001143762.1 NP_001137234.1 2 292
          NM_001970.4 NP_001961.1 2 307
          XM_005256509.2 XP_005256566.1 2 227
          XM_011523710.2 XP_011522012.1 2 224
          XM_011523711.2 XP_011522013.1 2 181
          XM_011523712.2 XP_011522014.1 2 244
          XM_011523713.2 XP_011522015.1 2 596
          XM_017024300.1 XP_016879789.1 2 286
          XM_017024301.1 XP_016879790.1 2 182
          GenBank mRNA AF087992.1 - 1 21
          AK222585.1 - 2 106
          AK292615.1 - 2 144
          AK303306.1 - 2 175
          AY129319.1 - 2 208
          AY129320.1 - 2 295
          AY129321.1 - 2 220
          AY129322.1 - 2 264
          BC000751.2 - 2 194 2961296
          BC001832.2 - 2 194 2961296
          BC030160.2 - 2 182 5022012
          BC080196.1 - 2 177 6528111
          BC085015.1 - 1 102 6575132
          BC107779.1 - 2 186 6495558
          BG479628.1 - 2 296
          CB111036.1 - 2 307
          M23419.1 - 1 113
          Gene Symbol: EIF5AL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001099692.1 NP_001093162.1 1 119
          GenBank mRNA BC070048.1 - 1 119

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001033681, XM_006246638, XM_006246637, XM_006246635, FQ220539, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001166591, NM_001166592, NM_001166589, NM_001166594, NM_181582, and others.

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA export from nucleus
          translational frameshifting
          protein export from nucleus
          nucleocytoplasmic transport
          apoptotic process
          positive regulation of cell proliferation
          peptidyl-lysine modification to peptidyl-hypusine
          positive regulation of translational elongation
          positive regulation of translational termination
          protein transport
          mRNA transport
          RNA binding
          translation elongation factor activity
          protein binding
          U6 snRNA binding
          ribosome binding
          poly(A) RNA binding
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0