Search
Search

ELF4| Gene Symbol: ELF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127197.1 | NP_001120669.1 | 4 | 625 | ||
| NM_001421.3 | NP_001412.1 | 4 | 707 | |||
| XM_005262389.3 | XP_005262446.1 | 4 | 1043 | |||
| XM_011531307.2 | XP_011529609.1 | 3 | 453 | |||
| XM_011531308.2 | XP_011529610.1 | 2 | 235 | |||
| GenBank mRNA | AF000670.1 | - | 3 | 424 | ||
| AK314972.1 | - | 4 | 624 | |||
| BC017194.2 | - | 4 | 590 | 3138355 | ||
| CN314421.1 | - | 4 | 622 | |||
| U32645.1 | - | 4 | 704 | |||