Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDK1 SIRNA Assays ›
          Gene Symbol:
          CDK1
          Gene Name:
          cyclin dependent kinase 1
          Gene Aliases:
          CDC2, CDC28A, P34CDC2
          Chromosome Location:
          Chr.10: 60778331 - 60794852 on Build GRCh38
          UniGene ID:
          Hs.732435
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s465
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CDK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001320918.1 NP_001307847.1 7 856
          NM_001786.4 NP_001777.1 7 844
          NM_033379.4 NP_203698.1 6 673
          XM_005270303.3 XP_005270360.1 7 926
          GenBank mRNA AK291939.1 - 7 781
          AW976143.1 - 6 588
          BC014563.1 - 6 715 2967728
          BC107750.1 - 7 830 6710672
          BT007004.1 - 6 702
          CR933728.1 - 6 3071
          D88357.1 - 5 558
          X05360.1 - 7 828
          Y00272.1 - 7 828

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00176469_m1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          regulation of transcription involved in G1/S transition of mitotic cell cycle
          G2/M transition of mitotic cell cycle
          activation of MAPK activity
          microtubule cytoskeleton organization
          DNA replication
          DNA repair
          protein complex assembly
          apoptotic process
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          mitotic nuclear division
          mitotic nuclear envelope disassembly
          mitotic G2 DNA damage checkpoint
          centrosome cycle
          pronuclear fusion
          cell aging
          response to toxic substance
          positive regulation of gene expression
          regulation of Schwann cell differentiation
          response to organic cyclic compound
          response to amine
          response to activity
          cell migration
          histone phosphorylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          chromosome condensation
          epithelial cell differentiation
          organ regeneration
          anaphase-promoting complex-dependent catabolic process
          positive regulation of protein import into nucleus, translocation
          protein localization to kinetochore
          response to drug
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of apoptotic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          response to ethanol
          positive regulation of DNA replication
          positive regulation of mitotic cell cycle
          regulation of embryonic development
          response to cadmium ion
          response to copper ion
          response to axon injury
          cell division
          negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
          positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
          regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
          ventricular cardiac muscle cell development
          positive regulation of cardiac muscle cell proliferation
          cellular response to hydrogen peroxide
          Golgi disassembly
          positive regulation of protein localization to nucleus
          protein kinase activity
          protein serine/threonine kinase activity
          cyclin-dependent protein serine/threonine kinase activity
          protein binding
          ATP binding
          RNA polymerase II carboxy-terminal domain kinase activity
          cyclin binding
          Hsp70 protein binding
          histone kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0