Search
Search

KIAA2012| Gene Symbol: KIAA2012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277372.2 | NP_001264301.2 | 2 | 592 | ||
| XM_006712172.2 | XP_006712235.1 | 2 | 600 | |||
| XM_011510467.2 | XP_011508769.1 | 2 | 600 | |||
| XM_011510468.2 | XP_011508770.1 | 2 | 600 | |||
| XM_017003111.1 | XP_016858600.1 | 2 | 600 | |||
| XM_017003112.1 | XP_016858601.1 | 2 | 600 | |||
| GenBank mRNA | AK096380.1 | - | 2 | 531 | ||
| AK295603.1 | - | 2 | 556 | |||
| AK297413.1 | - | 2 | 590 | |||