Search
Search

ADAMTS19| Gene Symbol: ADAMTS19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011543246.2 | XP_011541548.1 | 13 | 1653 | ||
| XM_011543247.2 | XP_011541549.1 | 13 | 1677 | |||
| XM_011543248.2 | XP_011541550.1 | 13 | 1797 | |||
| XM_011543249.2 | XP_011541551.1 | 8 | 1956 | |||
| XM_017009174.1 | XP_016864663.1 | 12 | 2412 | |||
| XM_017009175.1 | XP_016864664.1 | 9 | 2709 | |||
| XM_017009176.1 | XP_016864665.1 | 14 | 1825 | |||
| GenBank mRNA | AJ311904.1 | - | 14 | 2253 | ||
| AK122622.1 | - | 4 | 499 | |||
| AK128729.1 | - | 12 | 1468 | |||